missing translation for 'onlineSavingsMsg'
Learn More

Micro Particles Based on Polystyrene, Dark blue, Size: 100 m, MilliporeSigma™ Supelco™

$976.04

Specifications

Concentration 5% (solids)
Content And Storage 2°C to 8°C
Synonym Latex beads from PS, dark blue
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
111003774
View Documents Promotion Details
N/A 5 mL
Each for $976.04
Only null left
 
Description

Description

Specifications

Specifications

5% (solids)
2°C to 8°C
Latex beads from PS, dark blue
Videos
SDS
Documents

Documents