missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micro Particles Based on Polystyrene, Dark blue, Size: 100 m, MilliporeSigma™ Supelco™
Specifications
Specifications
| Concentration | 5% (solids) |
| Content And Storage | 2°C to 8°C |
| Quantity | 5 mL |
| Synonym | Latex beads from PS, dark blue |