missing translation for 'onlineSavingsMsg'
Learn More

Micro Particles Based on Polystyrene, Analytical Standard, Size: 15 m, MilliporeSigma™ Supelco™

Catalog No. 111004081
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004081 5 mL
111003814 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111004081 Supplier Merck Emd Millipore Supplier No. 749645MLF

Specifications

Density 1.05 g/cm3
Physical Form Particles
Concentration 10% (solids)
MDL Number MFCD00243243
Synonym Latex beads from PS
Quantity 5 mL
Grade Analytical Standard