missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microparticules à base de polyméthacrylate, norme analytique, taille : 50 m, MilliporeSigma™ Supelco™
Spécifications
Spécifications
| Physical Form | Particules |
| Concentration | 10% (solids) |
| MDL Number | MFCD00198073 |
| Synonym | 2-Propenoic acid,2-methyl homopolymer; 2-methyl-2-propenoic acid homopolymer; Microparticles based on PMAA; Microparticles based on poly(methacrylic acid); Polymethacrylate |
| Quantity | 10 mL |
| Grade | Norme analytique |