missing translation for 'onlineSavingsMsg'
Learn More

Micro Particles Based on Polymethacrylate, Analytical Standard, Size: 4 m, MilliporeSigma™ Supelco™

Catalog No. 111004254
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004254 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004254 Supplier Merck Emd Millipore Supplier No. 733715MLF

Specifications

Physical Form Particles
Concentration 10% (solids)
MDL Number MFCD00198073
Synonym 2-Propenoic acid,2-methyl homopolymer; 2-methyl-2-propenoic acid homopolymer; Microparticles based on PMAA; Microparticles based on poly(methacrylic acid); Polymethacrylate
Quantity 5 mL
Grade Analytical Standard