missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micro Particle Size Standard, Based on Polystyrene Monodisperse, Analytical Standard, Size: 30.0 m, MilliporeSigma™ Supelco™
Specifications
Specifications
| Physical Form | Particles |
| Concentration | 2% (solids) |
| MDL Number | MFCD00243243 |
| Synonym | Size standards |
| Quantity | 5 mL |
| Grade | Analytical Standard |
Coulter is a trademark of Beckman Coulter, Inc.