missing translation for 'onlineSavingsMsg'
Learn More

Micro Particle Size Standard, Based on Polystyrene Monodisperse, Analytical Standard, Size: 0.1 m, MilliporeSigma™ Supelco™

$366.70 - $612.29

Specifications

Physical Form Particles
Concentration 2% (solids)
MDL Number MFCD00243243
Synonym Size standards
Grade Analytical Standard
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
111004269
View Documents
Merck Emd Millipore
905175MLF
5 mL
Each for $366.70
Only null left
 
111003943
View Documents
Merck Emd Millipore
9051710MLF
10 mL
Each for $612.29
Only null left
 
Description

Description

Specifications

Specifications

Particles
MFCD00243243
Analytical Standard
2% (solids)
Size standards
Videos
SDS
Documents

Documents

Coulter is a trademark of Beckman Coulter, Inc.