missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micro Essential Labs Hydrion pH Buffers
Available in convenient envelopes or vials
$36.13 - $38.90
Specifications
| pH | 4.0 |
|---|---|
| Quantity | 5 Chemvelopes |
| Catalog Number | Mfr. No. | Quantity | pH | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | pH | Price | Quantity | ||||
|
14-853-151B
|
Micro Essentials Lab Inc
BUFFER40 |
5 Chemvelopes | 4.0 |
Pack of 5 Each for $38.90
|
|
||||
|
14-853-151C
|
Micro Essentials Lab Inc
BUFFER70 |
5 Chemvelopes | 7.0 |
Pack of 5 Each for $38.90
|
|
||||
|
14-853-151D
|
Micro Essentials Lab Inc
BUFFER90 |
5 Chemvelopes | 9.0 |
Pack of 5 Each for $36.13
|
|
||||
|
14-853-151A
|
Micro Essentials Lab Inc
BUFFER100 |
5 Chemvelopes | 10.0 |
Pack of 5 Each for $38.90
|
|
||||
Specifications
| 4.0 | |
| 5 Chemvelopes |