Learn More
Micro Essential Lab Hydrion™ Spectral Plastic pH Strips
Single color match — just dip and read
Supplier: Micro Essential Lab 9700
Description
- Economical — low cost per test
- Accurate — same high-quality pH test in plastic strip format
- Convenient — packed in handy flip-top vials
- Made in the USA
Chemical Identifiers
| Colorimetric chart | |
| pH Test Strip |
Specifications
| 5.5 to 8 | |
| 0.5 | |
| Each |
| To test urine and saliva, it offers an easy to read, single color match at every 0.2 to 0.3 interval from pH 5.5 to 8 | |
| Colorimetric chart | |
| pH Test Strip |