Learn More
Micro Essential Lab Hydrion™ Spectral Plastic pH Strips
Description
- Economical — low cost per test
- Accurate — same high-quality pH test in plastic strip format
- Convenient — packed in handy flip-top vials
- Made in the USA
Specifications
Specifications
| pH Range | 0 to 14 |
| Graduations | 1 |
| Includes | Colorimetric chart |
| Quantity | 100 Strips/Vial |
| Type | pH Test Strip |