Learn More
OHAUS Micro Centrifuge
Description
Frontier™ micro centrifuge for a wide range of essential lab applications.
Specifications
Specifications
| Capacity | 18 x 1.5/2 mL |
| Max. Speed | 14,000 rpm |
| Max. RCF | 16,000 x g |
| Refrigerated | No |
| Product Type | Centrifuge |