missing translation for 'onlineSavingsMsg'
Learn More

Miconazole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013606
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013606 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013606 Supplier Merck Emd Millipore Supplier No. PHR16181G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Miconazole is an antimycotic-imidazole derivative compound which is utilized for the management of topical fungal infections. It is typically effective against dermatophytes and Candida albicans, and also many Gram-positive germens.

Specifications

CAS 22916-47-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C18H14 Cl4N2 O
Synonym Miconazole; (+/-)-1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1 H-imidazole
Type Certified Reference Material
Quantity 1 g
Formula Weight 416.13
Grade Certified Reference Material