Learn More
MilliporeSigma™ Supelco™ Michel-Miller Swagelok™ adapter NPTF 1/16 in., PTFE
Use to make Swagelok™ tubing connections with Ace threaded glassware. One end is Ace-Thred, other is NPTF. Connects tubing to M-M columns. Adapters with No. 50 Ace-Thred fit Ace large columns.
$151.26 - $239.95
Specifications
Packaging Quantity 1/Pk.
Material PTFE
View More Specs
Products 2
Catalog Number Mfr. No. Thread Size Price Quantity  
Catalog Number Mfr. No. Thread Size Price Quantity  
111011024
View Documents
MilliporeSigma™ Supelco™
Z5072371EA
Ace-Thred No. 11
Each for $151.26
Only null left
111010624
View Documents
MilliporeSigma™ Supelco™
Z5073181EA
Ace-Thred No. 25
Each for $239.95
Only null left
Description
Description
Specifications
Specifications
1/Pk.
PTFE
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Swagelok is a registered trademark of Swagelok Company