Learn More
MilliporeSigma™ Supelco™ Michel-Miller Swagelok™ adapter NPTF 1/16 in., PTFE
Use to make Swagelok™ tubing connections with Ace threaded glassware. One end is Ace-Thred, other is NPTF. Connects tubing to M-M columns. Adapters with No. 50 Ace-Thred fit Ace large columns.
Supplier:  MilliporeSigma™ Supelco™ Z5073181EA
 View more versions of this product
Catalog No. 111010624
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
1/Pk.
Ace-Thred No. 25
PTFE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Swagelok is a registered trademark of Swagelok Company