Learn More
Invitrogen™ MHC Class II I-Ab Monoclonal Antibody (AF6-120.1), APC, eBioscience™, Invitrogen™
Description
Description: This AF6-120.1 monoclonal antibody reacts with the mouse MHC Class II I-Ab alloantigen of H-2b bearing mouse strains, including C57Bl/6 and 129. This cell surface molecule is involved in antigen presentation to T cells expressing CD3/TCR and CD4. The AF6-120.1 antibody crossreacts with H-2k and H-2u haplotypes. However, reactivity to other haplotypes (i.e., d, f, g7, p, q, r, and s) has not been observed. Applications Reported: This AF6-120.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This AF6-120.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | MHC Class II I-Ab |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | AF6-120.1 |
| Concentration | 0.2 mg/mL |
| Conjugate | APC |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | H2-Ab1 |
| Gene Accession No. | P01921, P06342, P06343, P06344, P06345, P06346, P14483 |
| Gene Alias | Abeta; AI845868; CELIAC1; DADB-249P12.2; H-2 class II histocompatibility antigen, A beta chain; H-2Ab; H2-Ab; H2-Ab1; H2Eb; H-2Eb; H2-iabeta; histocompatibility 2, class II antigen A, beta 1; HLA-DQB; Ia2; Ia-2; Ia4; Ia-4; IAb; I-Abeta; IDDM1; major histocompatibility complex class II beta chain; MGC163794; MGC163796; MHC class II antigen A beta; MHC class II H2-IA-beta-psi; response to metastatic cancers 1; Rmcs1 |
| Show More |
For Research Use Only.