missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MHC Class I Recombinant Rabbit Monoclonal Antibody (23GB6235)
GREENER_CHOICE

Catalog No. MA553004
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553004 100 μL
MA553005 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553004 Supplier Invitrogen™ Supplier No. MA553004
Only null left

Rabbit Recombinant Monoclonal Antibody

MHC Class I Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Like their mammalian counterparts, avian MHC Class I molecules, also known as B-F antigens, consist of a highly polymorphic alpha-chain noncovalently bound to the invariant beta2-microglobulin subunit. MHC Class I molecules are expressed on most nucleated cells where they present endogenously synthesized antigenic peptides to CD8+ T lymphocytes, which are usually cytotoxic T cells. The monoclonal antibody F21-2 also reacts with turkey MHC Class I. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.

Specifications

Antigen MHC Class I
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6235
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HLA-A
Gene Accession No. P04439
Gene Alias A29 protein; alpha chain 2; antigen presentation; antigen presentation; BFIV major; BF12 major; antigen presenting molecule; Aw-33; Aw-74; B12; BF; B-F; B-F antigen; G-B1; B-F histocompatibility F10 antigen; B-F major; B-F minor; BF1; B-F1; BF12; BF2; BFa1; BFa2; B-F-beta-IV; B-FI; BFIV; B-FIV; BFIV21; B-F-S01; B-F-S02; B-F-S03; B-F-S04; B-F-S05; B-F-S06; B-F-S07; B-F-S09; B-F-S11; B-F-S12; B-F-S13; B-F-S14; BFw-03; BFw-05; BFz-01; B-LB2; BOLA class I histocompatibility antigen, alpha chain BL3-7-like; chicken MHC class I antigen; class I histocompatibility antigen, F10 alpha chain; EA1.2; EA2.1; F10 alpha chain; HFE; HLA A; HLA class I antigen; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA class I histocompatibility antigen, alpha chain E; HLA class I histocompatibility antigen, E alpha chain; HLA locus; HLA-6.2; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLA-DQB1; HLA-DRB1; HLAE; HLA-E; Human leukocyte antigen A; leukocyte antigen class I-A; lymphocyte antigen; major class I glycoprotein; Major histocompatibility complex class I antigen BF2; major histocompatibility complex class I glycoprotein; major histocompatibility complex class I glycoprotein haplotype B21; major histocompatibility complex, class I, A; major histocompatibility complex, class I, E; major histocompatibility complex, class I, G; MCH class I antigen; MHC; MHC BF1 class I; MHC BF2 class I; MHC class 1 antigen; MHC class I; MHC class I alpha chain 1; MHC class I alpha chain 2; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen alpha chain; MHC class I antigen B-F heavy chain; MHC class I antigen B-F major heavy chain; MHC class I antigen B-F minor heavy chain; MHC class I antigen E; MHC class I antigen heavy chain; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen null protein; MHC class I antigene; MHC class I B-F heavy chain; MHC class I glycoprotein; MHC class I heavy chain B-F; MHC class I molecule; MHC class I-alpha; MHC class II antigen; MHC class1 antigen; MHC classI antigen; MHC HLA-E alpha-1; MHC HLA-E alpha-2.1; minor antigen; minor class I glycoprotein; OLA-I; OVAR; Ovar-I; ovar-MHCI-B564; ovar-MHCI-D5; ovar-MHCI-F12; put. F10 preprotein (AA -22 to 323); QA1; sHLA-E; Soluble HLA class I histocompatibility antigen, alpha chain E; truncated MHC class I antigen
Gene Symbols HLA-A
Host Species Rabbit
Immunogen A synthesized peptide derived from human MHC class I (250-300AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3105
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less