missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MHC Class I (H-2Kd) Monoclonal Antibody (SF1-1.1.1), APC-eFluor™ 780, eBioscience™, Invitrogen™

Catalog No. 47595780
Encompass
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47595780 25 μg
47595782 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47595780 Supplier Invitrogen™ Supplier No. 47595780

Mouse Monoclonal Antibody

This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.

Specifications

Antigen MHC Class I (H-2Kd)
Applications Flow Cytometry
Classification Monoclonal
Clone SF1-1.1.1
Concentration 0.2 mg/mL
Conjugate APC-eFluor 780
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene H2-K1
Gene Accession No. P01901, P03991, P04223, P14428
Gene Alias class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2-K1; histocompatibility 2, K1, K region; K-f; MGC184092; MGC7052; MHC class I antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-K transplantation antigen
Gene Symbols H2-K1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14972
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less