Learn More
Invitrogen™ mGluR6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA146253
Catalog No. PA146253
Only null left
Explore available promotions
Description
Description
This antibody reacts weakly on human brain tissue.
Glutamate receptors constitute the principal excitatory neurotransmitter receptors in brain. Two classes of glutamate receptors exist: Ionotropic receptors, and metabotropic receptors. Ionotropic glutamate receptors are oligomeric complexes of various subunits (GluR17, NMDA13, KAI2), which comprise ligand-gated calcium channels. Metabotropic glutamate receptors are G-protein coupled receptors, which when activated can activate phopholipases or adenylylcyclase, depending on the neuron.
Specifications
Specifications
mGluR6
Polyclonal
Unconjugated
GRM6
BC021919; CSNB1B; G protein-coupled receptor GRM6; G protein-coupled receptor, family C, group 1, member F; glutamate metabotropic receptor 6; glutamate receptor; glutamate receptor, metabotropic 6; Gm3; GPRC1F; Grm6; Metabotropic glutamate receptor 6; mGlu6; MGLUR6; nerg1; nob2; nob3; nob4
Rabbit
Antigen affinity chromatography
RUO
108072, 24419
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
P35349, Q5NCH9
GRM6
C-terminus of rat metabotropic Glutamate Receptor 6 (mGluR6).
100 μL
Primary
Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions