missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ mGluR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150194
Description
mGluR3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Glutamate is the main excitatory neurotransmitter in the brain. For many years it had been considered to act only on the ligand-gated receptor channels-termed NMDA, AMPA and kainite receptors that are involved in the fast excitatory synaptic transmission. Recently, glutamate has been shown to regulate enzymes producing second messengers via specific receptors coupled to G-proteins. These receptors are called metabotropic glutamate receptors. In expression systems, Group-I receptors stimulate phospholipase C as revealed by an increase in phosphoinositide turnover and calcium release from internal stores. Group-II and -III receptors are coupled to the inhibition of adenylyl cyclase. The Group-I receptors include mGluR1a and mGluR5. The Group-II receptors in include mGluR2 and mGluR3.Chemical Identifiers
| mGluR3 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P31422, Q14832, Q9QYS2 | |
| 100521916, 108069, 24416, 2913 | |
| Recombinant Metabotropic Glutamate Receptor 3 (GRM3) (Asp568-Phe819). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Pig | |
| GRM3 | |
| 0710001G23Rik; G protein-coupled receptor, family C, group 1, member C; GLUR3; glutamate metabotropic receptor 3; glutamate receptor; glutamate receptor, metabotropic 3; Gprc1c; GRM3; M mGlu3; Metabotropic glutamate receptor 3; mGlu3; mGluR3 | |
| GRM3 | |
| Primary | |
| Protein A |
Specifications
| mGluR3 | |
| Polyclonal | |
| Unconjugated | |
| GRM3 | |
| 0710001G23Rik; G protein-coupled receptor, family C, group 1, member C; GLUR3; glutamate metabotropic receptor 3; glutamate receptor; glutamate receptor, metabotropic 3; Gprc1c; GRM3; M mGlu3; Metabotropic glutamate receptor 3; mGlu3; mGluR3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100521916, 108069, 24416, 2913 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P31422, Q14832, Q9QYS2 | |
| GRM3 | |
| Recombinant Metabotropic Glutamate Receptor 3 (GRM3) (Asp568-Phe819). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Antibody |