missing translation for 'onlineSavingsMsg'
Learn More

Mexiletine Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America M20405G

 View more versions of this product

Catalog No. M20405G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Mexiletine Hydrochloride
1-4-5370
5 g
C11H18ClNO
mexiletine hydrochloride, mexitil, mexiletine hcl, mexiletene hydrochloride, 1-2,6-dimethylphenoxy-2-propanamine hydrochloride, 1-2,6-dimethylphenoxy propan-2-amine hydrochloride, 2-propanamine, 1-2,6-dimethylphenoxy-, hydrochloride, mexitil tn, 1-2,6-xylyloxy-2-aminopropane hydrochloride
CC1=C(C(=CC=C1)C)OCC(C)N.Cl
215.72
CHEBI:6917
≥98.0% (HPLC,N)
202°C
White-Yellow
Crystal-Powder at 20°C
MFCD00216024
NFEIBWMZVIVJLQ-UHFFFAOYNA-N
1-(2,6-dimethylphenoxy)propan-2-amine;hydrochloride
21467
215.72

Chemical Identifiers

1-4-5370
215.72
NFEIBWMZVIVJLQ-UHFFFAOYNA-N
21467
1-(2,6-dimethylphenoxy)propan-2-amine;hydrochloride
C11H18ClNO
MFCD00216024
mexiletine hydrochloride, mexitil, mexiletine hcl, mexiletene hydrochloride, 1-2,6-dimethylphenoxy-2-propanamine hydrochloride, 1-2,6-dimethylphenoxy propan-2-amine hydrochloride, 2-propanamine, 1-2,6-dimethylphenoxy-, hydrochloride, mexitil tn, 1-2,6-xylyloxy-2-aminopropane hydrochloride
CHEBI:6917
CC1=C(C(=CC=C1)C)OCC(C)N.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : KR9310000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions