missing translation for 'onlineSavingsMsg'
Learn More

Mexiletine Hydrochloride 98.0+%, TCI America™

Catalog No. M20401G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20401G 1 g
M20405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20401G Supplier TCI America Supplier No. M20401G

Chemical Identifiers

CAS 1-4-5370
Molecular Formula C11H18ClNO
Molecular Weight (g/mol) 215.72
MDL Number MFCD00216024
InChI Key NFEIBWMZVIVJLQ-UHFFFAOYNA-N
Synonym mexiletine hydrochloride, mexitil, mexiletine hcl, mexiletene hydrochloride, 1-2,6-dimethylphenoxy-2-propanamine hydrochloride, 1-2,6-dimethylphenoxy propan-2-amine hydrochloride, 2-propanamine, 1-2,6-dimethylphenoxy-, hydrochloride, mexitil tn, 1-2,6-xylyloxy-2-aminopropane hydrochloride
PubChem CID 21467
ChEBI CHEBI:6917
IUPAC Name 1-(2,6-dimethylphenoxy)propan-2-amine;hydrochloride
SMILES CC1=C(C(=CC=C1)C)OCC(C)N.Cl

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 215.72
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material Mexiletine Hydrochloride
RTECSNumber KR9310000
TSCA No