Learn More
Mettler Toledo™ Tweezers for Analytical and Calibration Weights
Catalog No. 01915378
Change view
Click to view available options
For Use With (Application):
1g-1kg Weights
1mg to 50g Weights
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
01915378 1mg to 50g Weights
01915379 1g-1kg Weights
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01915378 Supplier Mettler Toledo™ Supplier No. 15900
Accessories for Weights. Ergonomic Weight Handling - Ergonomic tweezers and weight forks are made of clean room suitable materials and allow professional weight handling and balance testing.
Constructed of metal with carbon reinforced nylon.
Specifications
Product Type Tweezers
Material Metal with Carbon Reinforced Nylon
For Use With (Application) 1mg to 50g Weights
Quantity Each