missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ SevenExcellence™ S500 pH/ISE Benchtop Meter
The S500 meter performs ion selective (ISE), pH, redox/ORP, and ISFET measurements. It can be expanded with additional modules to measure conductivity, resistivity, DO, salinity, TDS, and BOD. It has an intuitive touchscreen and the ability to create measurement shortcuts.
Supplier: METTLER TOLEDO™ 30046248
Description
- Ion selective (ISE), pH, redox/ORP, and ISFET meter than can be expanded with additional measurement modules to measure conductivity, resistivity, dissolved oxygen, salinity, TDS, and BOD
- Easily create measurement and calibration shortcuts
- 7 inch color touch screen with intuitive menu navigation
- Compatible with a wide range of electrodes for any application
- Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Meter connects to electrodes via BNC; S7 to BNC cables available
- Export data to a printer, Excel file, or LIMS system
- Automation available for measurements and cleaning in high-throughput applications
- Manage user permissions and access to ensure data integrity
- Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
- Optional comprehensive qualification package available including IQ/OQ/PQ
Specifications
| RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA | |
| ±0.1mV | |
| 13 Pre defined and 20 user defined standard | |
| Max. 5 points, 8 pre defined and 20 user defined buffer group | |
| 1 | |
| 0.001 μS/cm to 2000 mS/cm | |
| -2 to 20 | |
| 0.001 μS/cm to 2000 mS/cm | |
| 0.001 to 1 | |
| -30°C to +130°C | |
| Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese | |
| S500 pH/ISE Meter | |
| 0.001 to 999999mg/L ppm, 0.0001 to 100mol/L %, 0.001 to 100000mmol/L | |
| 0.001 to 1 | |
| 0.001 to 1 | |
| 0.001 to 1 | |
| 0.1°C |
| ±0.5% | |
| ±0.002 | |
| Max. 5 points, 8 pre defined and 20 user defined buffer group | |
| 20,000 | |
| TFT | |
| -2000 to 2000 mV | |
| 0.001 μS/cm to 2000 mS/cm | |
| 0.001 μS/cm to 2000 mS/cm | |
| 0.001/0.01/0.1 | |
| S500 pH/ISE meter, electrode holder, 2 buffer sachets each for pH 4.01, 7, 9.21, 10.01, EasyDirect pH activation key, test certificate | |
| 250 analyses (20.000 data points) | |
| pH/ISE Meter | |
| 0 to 999999mg/L | |
| 0.1/1mV | |
| 0.001 to 1 | |
| ±0.1°C |