missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
METTLER TOLEDO™ SevenExcellence™ S500 pH/ISE Benchtop Meter
The S500 meter performs ion selective (ISE), pH, redox/ORP, and ISFET measurements. It can be expanded with additional modules to measure conductivity, resistivity, DO, salinity, TDS, and BOD. It has a touchscreen, measurement shortcuts, and can export data to a printer, Excel file, or LIMS system.
Manufacturer: METTLER TOLEDO™ 30046251
Description
- Ion selective (ISE), pH, redox/ORP, and ISFET meter than can be expanded with additional measurement modules to measure conductivity, resistivity, dissolved oxygen, salinity, TDS, and BOD
- Easily create measurement and calibration shortcuts
- 7 inch color touch screen with intuitive menu navigation
- Compatible with a wide range of electrodes for any application
- Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Meter connects to electrodes via BNC; S7 to BNC cables available
- Export data to a printer, Excel file, or LIMS system
- Automation available for measurements and cleaning in high-throughput applications
- Manage user permissions and access to ensure data integrity
- Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
- Optional comprehensive qualification package available including IQ/OQ/PQ
Specifications
±0.5% | |
±0.002 | |
Max. 5 points, 8 pre defined and 20 user defined buffer group | |
20,000 | |
Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese | |
S500 Kit - Fluoride Measurement | |
0 to 999999mg/L | |
0.001 to 1 | |
0.001 to 1 | |
±0.1°C | |
Single-channel | |
RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA | |
0.001μS/cm to 2000mS/cm | |
0.001μS/cm to 2000mS/cm | |
-2 to 20 | |
0.001/0.01/0.1 |
±0.1mV | |
13 Pre defined and 20 user defined standard | |
Max. 5 points, 8 pre defined and 20 user defined buffer group | |
S500 pH/ISE meter, perfectION comb F ion selective electrode (1.2 m cable), electrode holder, 2 buffer sachets each for pH 4.01, 7, 9.21, 10.01, EasyDirect pH activation key, test certificate, pH measurement guide | |
250 analyses (20.000 data points) | |
0.001 to 999999mg/L ppm, 0.0001 to 100mol/L %, 0.001 to 100000mmol/L | |
0.001 to 1 | |
0.001 to 1 | |
0.1/1mV | |
0.1°C | |
TFT | |
0.001 μS/cm to 2000 mS/cm | |
0.001μS/cm to 2000mS/cm | |
-2000 to 2000mV | |
0.001 to 1 | |
-30°C to +130°C |
Promotions
-
Earn Up to 5X the Fisher Scientific Buck$ with Purchase
Bundle your purchase of qualifying METTLER TOLEDO balances or analytical instruments to earn more.: 06/30/21