missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ SevenExcellence™ S470 pH/Conductivity Benchtop Meter
The S470 meter measures pH, conductivity, resistivity, redox/ORP, TDS, salinity, and ISFET. It can be expanded with additional modules to measure dissolved oxygen, BOD, and ISE. It has an intuitive touchscreen, measurement shortcuts, and can export data to a printer, Excel file, or LIMS system.
$7971.30 - $7971.30
Specifications
| Outputs | RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA |
|---|---|
| Accuracy (Conductivity) | ±0.5% |
| Accuracy (mV) | ±0.1mV |
| Accuracy (pH) | ±0.002 |
| Calibration Points (Conductivity) | 13 Pre defined and 20 user defined standard |
| Catalog Number | Mfr. No. | Model | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Model | Price | Quantity | |||||
|
01-910-915
|
METTLER TOLEDO™
64091338 |
S470-Bio, InLab Routine Pro |
Each for $7,971.30
|
|
|||||
Description
- pH, conductivity, resistivity, redox/ORP, TDS, salinity, and ISFET meter than can be expanded with additional measurement modules to measure dissolved oxygen, ISE, and BOD
- Easily create measurement and calibration shortcuts
- 7 inch color touch screen with intuitive menu navigation
- Compatible with a wide range of electrodes for any application
- Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Meter connects to electrodes via BNC; S7 to BNC cables available
- Export data to a printer, Excel file, or LIMS system
- Automation available for measurements and cleaning in high-throughput applications
- Manage user permissions and access to ensure data integrity
- Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
- Optional comprehensive qualification package available including IQ/OQ/PQ
Specifications
| RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA | |
| ±0.1mV | |
| 13 Pre defined and 20 user defined standard | |
| Max. 5 points, 8 pre defined and 20 user defined buffer group | |
| TFT | |
| -2000 to 2000 mV | |
| 0.01 to 100 Mohm/cm | |
| 0.001 mg/L to 1000 g/L | |
| 0.001/0.01/0.1 | |
| 0 to 999999mg/L | |
| 0.1/1mV | |
| 0.001 to 1 | |
| ±0.1°C |
| ±0.5% | |
| ±0.002 | |
| Max. 5 points, 8 pre defined and 20 user defined buffer group | |
| 20,000 | |
| 0.001 μS/cm to 2000 mS/cm | |
| -2 to 20 | |
| 0 to 80 ppt | |
| 0.001 to 1 | |
| -30°C to +130°C | |
| 0.001μS/cm to 2000mS/cm | |
| 0.001 to 1 | |
| 0.001 to 1 | |
| 0.1°C |