missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ S479 SevenExcellence™ pH/Conductivity/DO Benchtop Meter Kit

Catalog No. 01910926
Change view
Click to view available options
Channels:
Multiple
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Channels
01910926 Multiple
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01910926 Supplier METTLER TOLEDO™ Supplier No. 30092988

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

SevenExcellence pH/mV/Conductivity/DO triple channel meter (instrument with a pH/mV, conductivity and DO expansion unit); as well as an InLab™ ExpertPro-ISM (pH), InLab 731-ISM (conductivity) and InLab OptiOx (digital DO) sensor; pH buffers; conductivity standards; and zero-oxygen calibration tablets; guides to pH, conductivity, and DO measurement; uPlace™ electrode holder, semi-transparent cover, operating instructions, installation, LabX™ direct pH PC software, declaration of conformity and test certificate

The S479 meter kit gives you everything you need to start measuring pH, conductivity, dissolved oxygen, BOD, resistivity, salinity, TDS, Redox/ORP, and ISFET all on one meter. It has an intuitive touchscreen, measurement shortcuts, and can export data to a printer, Excel file, or LIMS system.

S479 meter kit containing SevenExcellence meter, electrode, and accessories to perform pH, conductivity, dissolved oxygen, resistivity, salinity, TDS, BOD, Redox/ORP, and ISFET measurements

  • The SevenExcellence can be expanded with additional modules to measure pH, conductivity, resistivity, ISE, salinity, TDS, Redox/ORP, and ISFET
  • Easily create measurement and calibration shortcuts
  • 7 inch color touch screen with intuitive menu navigation
  • Compatible with a wide range of electrodes for any application
  • Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Meter connects to electrodes via BNC; S7 to BNC cables available
  • Export data to a printer, Excel file, or LIMS system
  • Automation available for measurements and cleaning in high-throughput applications
  • Manage user permissions and access to ensure data integrity
  • Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
  • Optional comprehensive qualification package available including IQ/OQ/PQ
  • Preventative maintenance and service available through METTLER TOLEDO Service

Specifications

Description SevenExcellence pH/Cond/DO meter S479-K
Channels Multiple
Display Type Color TFT
IP Rating IP54
Measurement Modes pH, Conductivity, Dissolved Oxygen
Percent Saturation Range (Polarographic DO) 0 to 600%
Range (mV) -2000 to 2000 mV
Range (pH) -2.000 to 20.000
Range (Resistivity) 0.01 to 100 MOhm-cm
Range (Salinity) 0 to 80 ppt
Range (TDS) 0.001 to 1000 g/L
Resolution (ISE) 0.001 mg/L
Resolution (pH) 0.001
Temperature Range (Metric) -30°C to +130°C
Includes S479 pH/Conductivity/DO meter, InLab Expert Pro-ISM pH electrode (1.2 m cable), InLab 731-ISM conductivity electrode, InLab OptiOx Optical dissolved oxygen electrode (1.8 m cable), InLab OptiOx calibration tube, 2 buffer sachets each for pH 4.01, 7, 9.21, and 10.01, 2 buffer sachets each for pH 1413 mS/cm and 12.88 mS/cm), EasyDirect pH activation key, test certificate, pH measurement guide, conductivity/DO measurement guide
Languages Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese
Memory 250 analyses (20.000 data points)
Model S479 pH/Conductivity/DO Kit
Product Type S479 SevenExcellence pH/Conductivity/DO Meter Kit
Show More Show Less