missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mettler Toledo™ Moisture Analyzer Sample Pans
Mettler Toledo sample pans are the only sample pans that are officially supported by Mettler Toledo Moisture Analyzers for optimal use and guaranteed performance.
Manufacturer: Mettler Toledo™ 13954
Description
Choice of washable, reusable stainless chrome-nickel steel pans for halogen moisture analyzers, or aluminum or stainless-steel pans for all Mettler Toledo moisture analyzers.
- 3.75 in. (96mm) in diameter
Specifications
15 mm | |
Reusable Pans - High edges (15mm) | |
Steel | |
All Mettler Toledo Moisture Analyzers |
Round | |
88.9 mm | |
Moisture Pan | |
50 g |