Learn More
Mettler Toledo™ LabX™ Balance Express Software
Catalog No. p-4377413
Change view
Click to view available options
For Use With (Equipment):
Excellence Series Balance
Excellence XP or XS Balance
Includes:
Balance Express Starter Pack: For installation on
Express Edition License: for 1 Excellence XP or XS
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Includes
01909386 Excellence Series Balance Balance Express Starter Pack: For installation on
01909387 Excellence XP or XS Balance Express Edition License: for 1 Excellence XP or XS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01909386 Supplier Mettler Toledo™ Supplier No. 11153120
Only null left
Weighs different classes of material with improved documentation allowing electronic integration of weights and sample IDs with other software systems within lab.
  • Express edition
  • Connect one instrument with option to up to 3 balances
  • Graphical method editor supports any weighing application
  • Built in method library for quick weighing method development
  • Comprehensive and customizable report editor
LabX seamlessly integrates with third-party systems such as LIMS, ELN, SAP and CDS
Specifications
For Use With (Equipment) Excellence Series Balance
For Use With (Application) Weighs different classes of material with improved
Includes Balance Express Starter Pack: For installation on
Type Balance Software