missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ ISA Solutions
Solutions for perfectION™ combined ion selective electrodes
Supplier: Mettler Toledo™ 51344760
Description
Solutions (ionic-strength adjustment buffers)
- Ion Electrolyte B (for chloride, copper, iodide, silver/sulfide ISE's), 475mL
- ISA for calcium electrode, 475mL
- ISA for potassium electrode, 475mL
- ISA for nitrate electrode, 475mL
- Nitrate ISS (for suppressing interference), 475mL
- Fluoride TISAB II with CDTA, 3790mL
- Fluoride TISAB III with CDTA (concentrate), 475mL
Specifications
| perfectION ISA solid state ISE | |
| ISA Solution | |
| 475 mL |
| ISE Combination Electrode | |
| Ion Electrolyte B (Chloride, copper, iodide, silver) | |
| 475 mL |