missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ ISA Solutions
Description
Solutions (ionic-strength adjustment buffers)
- Ion Electrolyte B (for chloride, copper, iodide, silver/sulfide ISE's), 475mL
- ISA for calcium electrode, 475mL
- ISA for potassium electrode, 475mL
- ISA for nitrate electrode, 475mL
- Nitrate ISS (for suppressing interference), 475mL
- Fluoride TISAB II with CDTA, 3790mL
- Fluoride TISAB III with CDTA (concentrate), 475mL
Specifications
Specifications
| Description | perfectION ISA solid state ISE |
| For Use With (Equipment) | ISE Combination Electrode |
| Type | ISA Solution |
| Filling Solution Type | Ion Electrolyte B (Chloride, copper, iodide, silver) |
| Packaging | 475 mL |
| Quantity | 475 mL |