missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ ISA Solutions

Solutions for perfectION™ combined ion selective electrodes

Supplier:  Mettler Toledo™ 51344762

 View more versions of this product

Catalog No. 01928352


Only null left

Description

Description

Solutions (ionic-strength adjustment buffers)

  • Ion Electrolyte B (for chloride, copper, iodide, silver/sulfide ISE's), 475mL
  • ISA for calcium electrode, 475mL
  • ISA for potassium electrode, 475mL
  • ISA for nitrate electrode, 475mL
  • Nitrate ISS (for suppressing interference), 475mL
  • Fluoride TISAB II with CDTA, 3790mL
  • Fluoride TISAB III with CDTA (concentrate), 475mL

Specifications

Specifications

perfectION potassium ISA
ISA Solution
475 mL
Potassium Electrode
Potassium
475 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions