missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ F2 Adjusting Cavity

Catalog No. 01911782
Change view
Click to view available options
F2 Adjusting Cavity
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01911782 F2 Adjusting Cavity
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01911782 Supplier Mettler Toledo™ Supplier No. 30406432

Basic Line Weights - Design with adjusting cavity allows cost and time saving production processes. These favorably priced weights are perfect for routine testing. High quality, stainless steel for all weights offers longer life-span.

  • For use with PL601-S Portable Balance
  • Weight: 500g
  • Class F2
  • AC in plastic box
  • Includes Calibration Certificate
  • Weight calibration performed by robots - substantial investments in cutting-edge robot technology combined with decades of experience have opened up a new field in weight calibration.

Specifications

Product Type F2 Adjusting Cavity
Material Stainless Steel
For Use With (Equipment) PL601-S Portable Balance
Includes With certificate