missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standards for METTLER TOLEDO Conductivity Probes, METTLER TOLEDO™
Recommended NIST traceable solutions for calibrating or verifying METTLER TOLEDO conductivity electrodes.
Supplier: METTLER TOLEDO 51302050
Description
- For daily verification: to monitor health of electrode and ensure measurement accuracy
- For initial calibration of nominal cell constant electrodes
- Be sure to screw cap tightly immediately after use (sachets minimize the chance of error due to environmental interaction)
- Conductivity standards subject to change values when exposed to environment; water evaporation raises conductivity values
Specifications
| Potassium Chloride Solution | |
| 7447-40-7,7732-18-5 | |
| 12.88 mS/cm | |
| Green/White | |
| 0°C | |
| Conductivity Electrodes | |
| Conductivity Standard | |
| 20 mL |
| 30 x 20 mL | |
| Yes | |
| KCl | |
| Approx. 6 | |
| 100°C | |
| Conductivity Standard | |
| Calibration of Conductivity Meter | |
| Laboratory |