Learn More
Mettler Toledo™ CarePacs™: Customized Third Weight
Catalog No. p-3577244
Change view
Click to view available options
Weight (Metric):
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
1 g
10 g
20 g
50 g
100 g
Classification:
ASTM Class 1
OIML Class E2
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric) Classification
01912117 2 mg ASTM Class 1
01912119 10 mg ASTM Class 1
01912120 20 mg ASTM Class 1
01912121 50 mg ASTM Class 1
01912122 100 mg ASTM Class 1
01912125 1 g ASTM Class 1
01912134 5 mg OIML Class E2
01912138 100 mg OIML Class E2
01912139 200 mg OIML Class E2
01912141 1 g OIML Class E2
01912144 10 g OIML Class E2
01912145 20 g OIML Class E2
01912146 50 g OIML Class E2
01912147 100 g OIML Class E2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 01912117 Supplier Mettler Toledo™ Supplier No. 11123145
Only null left
Third weight for customer-specific testing (e.g., minimum weight determination)
  • Optional customized third weight for Mettler Toledo CarePacs
  • ASTM or OIML certified
Specifications
Product Type Customized 3rd Weight
Certifications/Compliance ISO9001, ISO14001 environmental standard
Classification ASTM Class 1
Material Stainless Steel
Weight (Metric) 2 mg
Mass 2 mg
Quantity Each
Packaging Quantity 1