missing translation for 'onlineSavingsMsg'
Learn More

Mettler-Toledo™ Calibration substance ME 51143093, Vanillin traceable to primary standards (LGC), MilliporeSigma™ Supelco™

Catalog No. 111012766
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012766 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012766 Supplier Merck Emd Millipore Supplier No. 776345G
Only null left
Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care

Specifications

Product Line Mettler-Toledo
CAS 121-33-5
Vapor Pressure >0.01 mmHg (25°C)
Vapor Density 5.3 (vs. air)
Flash Point 153°C - Closed Cup
Physical Form Neat
Linear Formula 4-(HO)C6H3-3-(OCH3)CHO
MDL Number MFCD00006942
Melting Point 81°C to 83°C
Boiling Point 170°C (15 mmHg)
Synonym Vanillin; 4-Hydroxy-3-methoxybenzaldehyde; Vanillic aldehyde
Type Analytical Standard
Quantity 5 g
Formula Weight 152.15
Grade Analytical Standard
Show More Show Less

Mettler-Toledo is a registered trademark of Mettler-Toledo, Inc.