missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Balance Printer Accessories: Printer Paper

Catalog No. 0191078
Change view
Click to view available options
Quantity:
5 Rolls/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
0191078 5 Rolls/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0191078 Supplier Mettler Toledo™ Supplier No. 72456

Printer paper for use with Mettler Toledo™ balances.

  • Color: White
  • Five rolls per pack
  • For use with Mettler Toledo balances: LC-P45, LC-P43, HA-P43, HR-73P

Specifications

For Use With (Equipment) LC-P45, LC-P43, HA-P43, HR-73P Printer
Quantity 5 Rolls/Pk.
Type Printer Paper