missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metrizoic Acid 98.0+%, TCI America™
Supplier: TCI America M13635G
Specifications
| Metrizoic Acid | |
| White | |
| MFCD00867964 | |
| 3-Acetamido-5-(N-methylacetamido)-2,4,6-triiodobenzoic Acid | |
| CN(C(C)=O)C1=C(I)C(NC(C)=O)=C(I)C(C(O)=O)=C1I | |
| 627.94 | |
| CHEBI:34847 | |
| ≥98.0% (T,HPLC) |
| 1949-45-7 | |
| C12H11I3N2O4 | |
| 5 g | |
| GGGDNPWHMNJRFN-UHFFFAOYSA-N | |
| 3-acetamido-2,4,6-triiodo-5-(N-methylacetamido)benzoic acid | |
| 2528 | |
| 627.94 | |
| Crystalline Powder |
Chemical Identifiers
| 1949-45-7 | |
| 627.94 | |
| GGGDNPWHMNJRFN-UHFFFAOYSA-N | |
| 2528 | |
| 3-acetamido-2,4,6-triiodo-5-(N-methylacetamido)benzoic acid |
| C12H11I3N2O4 | |
| MFCD00867964 | |
| 3-Acetamido-5-(N-methylacetamido)-2,4,6-triiodobenzoic Acid | |
| CHEBI:34847 | |
| CN(C(C)=O)C1=C(I)C(NC(C)=O)=C(I)C(C(O)=O)=C1I |
Safety and Handling
RTECSNumber : DG0887300
TSCA : No