missing translation for 'onlineSavingsMsg'
Learn More

Medisca Metoprolol Tartrate USP

Catalog No. 14186092
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186092 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14186092 Supplier Medisca Supplier No. 057808

Metoprolol Tartrate USP is a white, crystalline powder.

Metoprolol Tartrate USP is a white, crystalline powder.

Specifications

Chemical Name or Material Metoprolol Tartrate USP
Packaging Jar, 2.8L, White, HDPE, Round, 110-400, TE Pano Neck
CAS 56392-17-7
Type Drug Standard
Molecular Weight (g/mol) 684.8
Quantity 500 g
Percent Purity 98 to 102 %
Grade USP