missing translation for 'onlineSavingsMsg'
Learn More

Metoprolol Tartrate 98.0+%, TCI America™

Catalog No. p-7133291
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25555G 5 g
M255525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25555G Supplier TCI America Supplier No. M25555G
Only null left

Chemical Identifiers

CAS 56392-17-7
Molecular Formula C34H56N2O12
Molecular Weight (g/mol) 684.824
MDL Number MFCD00056257
InChI Key YGULWPYYGQCFMP-NPHUUBOGSA-N
Synonym y-metoprolol +-tartrate, ?-metoprolol +-tartrate, --tartaric acid; bis metoprolol, 1-isopropylamino-3-p-beta-methoxyethyl phenoxy-2-propanol +-tartrate salt, 1-isopropylamino-3-4-2-methoxyethyl phenoxy propan-2-ol 2,3-dihydroxysuccinate 2
PubChem CID 11957594
IUPAC Name (2R,3S)-2,3-dihydroxybutanedioic acid;1-[4-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
SMILES CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.CC(C)NCC(COC1=CC=C(C=C1)CCOC)O.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 123°C
Color White
Quantity 5 g
Formula Weight 342.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Metoprolol Tartrate
RTECSNumber UB7450100
TSCA No