missing translation for 'onlineSavingsMsg'
Learn More

Metoclopramide hydrochloride

Catalog No. AAJ6154509
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6154509 10 g
AAJ6154514 25 g
AAJ6154522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6154509 Supplier Thermo Scientific Chemicals Supplier No. J6154509

5-HT3 serotonin antagonist; D2 dopamine antagonist | CAS: 54143-57-6 | C14H25Cl2N3O3 | 354.27 g/mol

Metoclopramide hydrochloride is used as a digestive system drugs. It is used for anti-emetic, used for digestive dysfunction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metoclopramide hydrochloride is used as a digestive system drugs. It is used for anti-emetic, used for digestive dysfunction.

Solubility
It is very soluble in water, easily soluble in ethyl alcohol, slightly soluble in dichloromethane.

Notes
Hygroscopic, Refrigerator, Under Inert atmosphere. Incompatible to strong alkalis, strong oxidizing agents.

Chemical Identifiers

CAS 54143-57-6
Molecular Formula C14H25Cl2N3O3
Molecular Weight (g/mol) 354.27
MDL Number MFCD00150634
InChI Key KJBLQGHJOCAOJP-UHFFFAOYSA-N
PubChem CID 441347
ChEBI CHEBI:6899
IUPAC Name 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrate hydrochloride
SMILES O.Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC

Specifications

Color White to Yellow
Quantity 10 g
Sensitivity Light sensitive
Merck Index 14,6143
Solubility Information It is very soluble in water,easily soluble in ethyl alcohol,slightly soluble in dichloromethane.
Formula Weight 336.26
Physical Form Crystalline Solid
Chemical Name or Material Metoclopramide hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only