missing translation for 'onlineSavingsMsg'
Learn More

Metoclopramide Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002472
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002472 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002472 Supplier Merck Emd Millipore Supplier No. PHR11321G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Metoclopramide Hydrochloride is an antiemetic drug and is generally used for the treatment of gastrointestinal disorders. It can also exhibit sedative, extrapyrimidal, and prolactin secretion stimulation actions.

Specifications

CAS 54143-57-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Type Certified Reference Material
Quantity 1 g
Grade Certified Reference Material