missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium Iodide 98.0+%, TCI America™

Catalog No. p-7132079
Change view
Click to view available options
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
M025325G 25 g
M0253100G 100 g
M0253500G 500 g
3 options

Catalog No. M025325G

Supplier: TCI America M025325G

Only null left

Chemical Identifiers

CAS 2065-66-9
Molecular Formula C19H18IP
Molecular Weight (g/mol) 404.23
MDL Number MFCD00066175
InChI Key JNMIXMFEVJHFNY-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium iodide, methyl triphenylphosphonium iodide, methyltriphenylphosphanium iodide, methyl triphenyl phosphonium iodide, phosphonium, methyltriphenyl-, iodide, methyl triphenylphosphoniumiodide, methyl triphenyl phosphanium iodide, triphenylmethyl phosphonium iodide, pubchem15730
PubChem CID 638159
IUPAC Name methyltriphenylphosphanium iodide
SMILES [I-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 185°C
Color White
Quantity 25 g
Formula Weight 404.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methyltriphenylphosphonium Iodide