missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium iodide, 98%

Catalog No. AAA1564422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1564422 100 g
AAA1564414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1564422 Supplier Thermo Scientific Chemicals Supplier No. A1564422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2065-66-9 | C19H18IP | 404.23 g/mol

Methyltriphenylphosphonium iodide is as a reactant for synthesis of triphenylamine-based dyes and polyolefinic aromatic molecules with pyrene for use in dye-sensitized solar cells. Used as ligand in coupling reaction. It is also used as a precursor to a Wittig reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltriphenylphosphonium iodide is as a reactant for synthesis of triphenylamine-based dyes and polyolefinic aromatic molecules with pyrene for use in dye-sensitized solar cells. Used as ligand in coupling reaction. It is also used as a precursor to a Wittig reagent.

Solubility
Soluble in water, acetone, dichloromethane , methanol.

Notes
Light sensitive and hygroscopic. Store in dark. Protect from humidity and water.

Chemical Identifiers

CAS 2065-66-9
Molecular Formula C19H18IP
Molecular Weight (g/mol) 404.23
MDL Number MFCD00066175
InChI Key JNMIXMFEVJHFNY-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium iodide, methyl triphenylphosphonium iodide, methyltriphenylphosphanium iodide, methyl triphenyl phosphonium iodide, phosphonium, methyltriphenyl-, iodide, methyl triphenylphosphoniumiodide, methyl triphenyl phosphanium iodide, triphenylmethyl phosphonium iodide, pubchem15730
PubChem CID 638159
IUPAC Name methyl(triphenyl)phosphanium;iodide
SMILES [I-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 185°C to 188°C
Odor Odorless
Quantity 100 g
UN Number UN3464
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water,acetone,dichloromethane ,methanol.
Formula Weight 404.23
Percent Purity 98%
Chemical Name or Material Methyltriphenylphosphonium iodide
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
EINECSNumber 218-178-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only