missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium iodide, 97%

Catalog No. AC224030250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC224030250 25 g Plastic bottle
AC224031000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC224030250 Supplier Thermo Scientific Chemicals Supplier No. 224030250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2065-66-9 | C19H18IP | 404.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 2065-66-9
Molecular Formula C19H18IP
Molecular Weight (g/mol) 404.23
MDL Number MFCD00066175
InChI Key JNMIXMFEVJHFNY-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium iodide, methyl triphenylphosphonium iodide, methyltriphenylphosphanium iodide, methyl triphenyl phosphonium iodide, phosphonium, methyltriphenyl-, iodide, methyl triphenylphosphoniumiodide, methyl triphenyl phosphanium iodide, triphenylmethyl phosphonium iodide, pubchem15730
PubChem CID 638159
SMILES [I-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 185.0°C to 187.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula CH3P(C6H5)3I
Quantity 25 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,ethanol,practically insoluble in aromatic hydrocarbons
IUPAC Name methyltriphenylphosphanium iodide
Formula Weight 404.23
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Methyltriphenylphosphonium iodide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-178-5

RUO – Research Use Only