missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium bromide, 98+%

Catalog No. AAA1587818
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1587818 50 g
AAA1587830 250 g
AAA158780B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1587818 Supplier Thermo Scientific Chemicals Supplier No. A1587818

CAS: 1779-49-3 | C19H18BrP | 357.23 g/mol

Methyltriphenylphosphonium bromide is used for methylenation through the Wittig reaction. It is utilized in the synthesis of an enyne and 9-isopropenyl -phenanthrene by using sodium amide as reagent. It is a lipophilic molecule and used to deliver molecules to specific cell components. Further, it acts as an antineoplastic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltriphenylphosphonium bromide is used for methylenation through the Wittig reaction. It is utilized in the synthesis of an enyne and 9-isopropenyl -phenanthrene by using sodium amide as reagent. It is a lipophilic molecule and used to deliver molecules to specific cell components. Further, it acts as an antineoplastic agent.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1779-49-3
Molecular Formula C19H18BrP
Molecular Weight (g/mol) 357.23
MDL Number MFCD00011804
InChI Key LSEFCHWGJNHZNT-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium bromide, methyltriphenylphosphanium bromide, triphenylmethylphosphonium bromide, methyl triphenylphosphonium bromide, methyl triphenyl phosphonium bromide, phosphonium, methyltriphenyl-, bromide, methyltriphenylphosphoniumbromide, methyl-triphenyl-phosphonium bromide
PubChem CID 74505
IUPAC Name methyl(triphenyl)phosphanium;bromide
SMILES [Br-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 231°C to 234°C
Assay Percent Range ≥98%
Quantity 50 g
UN Number UN2811
Beilstein 3599467
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 357.24
Percent Purity ≥98%
Chemical Name or Material Methyltriphenylphosphonium bromide
Hazard Category H301-H335-H500
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 217-218-9
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only