Learn More
Methyltrichlorosilane, 97%
Description
Methyltrichlorosilane is used in production of methyl silicone resins, its vapor reacts with water on surfaces to give a thin layer of methylpolysiloxane which make it a water-repellent film. A combination of methyltrichlorosilane and sodium iodide can be used to cleave carbon-oxygen bonds such as methyl ethers. It is used as a precursor for forming various cross-linked siloxane polymers. Trichloromethylsilane is the starting material for the production of pure silicon for manufacture of semiconductors and optical fibers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyltrichlorosilane is used in production of methyl silicone resins, its vapor reacts with water on surfaces to give a thin layer of methylpolysiloxane which make it a water-repellent film. A combination of methyltrichlorosilane and sodium iodide can be used to cleave carbon-oxygen bonds such as methyl ethers. It is used as a precursor for forming various cross-linked siloxane polymers. Trichloromethylsilane is the starting material for the production of pure silicon for manufacture of semiconductors and optical fibers.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture.
Specifications
Specifications
| Boiling Point | 64°C to 66°C |
| Melting Point | -78°C |
| Quantity | 500 g |
| Flash Point | 8°C (46°F) |
| Odor | Pungent |
| Refractive Index | 1.4115 |
| Sensitivity | Moisture sensitive |
| Density | 1.27 |
| UN Number | UN1250 |
| Beilstein | 1361381 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302+H312-H314-H318-H331 |
| Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P331-P362-P372-P374-P380-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYLTRICHLOROSILANE |
| EINECSNumber | 200-902-6 |
| RTECSNumber | VV4550000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only