missing translation for 'onlineSavingsMsg'
Learn More

Methyltri-n-octylammonium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. M16605G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M16605G Supplier TCI America Supplier No. M16605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 375395-33-8
Molecular Formula C27H54F6N2O4S2
Molecular Weight (g/mol) 648.849
InChI Key LAGQNGWYNLUQRI-UHFFFAOYSA-N
PubChem CID 16211009
IUPAC Name bis(trifluoromethylsulfonyl)azanide;methyl(trioctyl)azanium
SMILES CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point -75°C
Quantity 5 g
Formula Weight 648.85
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material Methyltri-n-octylammonium Bis(trifluoromethanesulfonyl)imide