accessibility menu, dialog, popup

UserName

CH3NCH3CH3CH3FFFSOONSOOFFF

CAS RN 375395-33-8

IUPAC Name: methyltrioctylazanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: trioctylmethylammonium bis(trifluoromethylsulfonyl)imide
Molecular Weight (g/mol): 648.85
Molecular Formula: C27H54F6N2O4S2
InChi Key: LAGQNGWYNLUQRI-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results