missing translation for 'onlineSavingsMsg'
Learn More

(Methylthio)acetonitrile 99.0+%, TCI America™

Catalog No. M094125G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M094125G 25 g
M094110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M094125G Supplier TCI America Supplier No. M094125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35120-10-6
Molecular Formula C3H5NS
Molecular Weight (g/mol) 87.14
MDL Number MFCD00001925
InChI Key ZRIGDBVSVFSVLL-UHFFFAOYSA-N
Synonym methylthio acetonitrile, methylthioacetonitrile, methylthio-acetonitrile, 2-methylsulfanyl acetonitrile, acetonitrile, methylthio, ch3sch2cn, acmc-1afh2, methylsulfanyl acetonitrile #, inchi=1/c3h5ns/c1-5-3-2-4/h3h2,1h
PubChem CID 99295
IUPAC Name 2-methylsulfanylacetonitrile
SMILES CSCC#N

Specifications

Color Yellow
Boiling Point 60°C
Quantity 25 g
UN Number 1760
Formula Weight 87.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material (Methylthio)acetonitrile
TSCA No
Recommended Storage Refrigerator