missing translation for 'onlineSavingsMsg'
Learn More

(Methylthio)acetonitrile, 99%

Catalog No. AAL0306622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0306622 100 g
AAL0306606 5 g
AAL0306614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0306622 Supplier Thermo Scientific Chemicals Supplier No. L0306622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35120-10-6 | C3H5NS | 87.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35120-10-6
Molecular Formula C3H5NS
Molecular Weight (g/mol) 87.14
MDL Number MFCD00001925
InChI Key ZRIGDBVSVFSVLL-UHFFFAOYSA-N
Synonym methylthio acetonitrile, methylthioacetonitrile, methylthio-acetonitrile, 2-methylsulfanyl acetonitrile, acetonitrile, methylthio, ch3sch2cn, acmc-1afh2, methylsulfanyl acetonitrile #, inchi=1/c3h5ns/c1-5-3-2-4/h3h2,1h
PubChem CID 99295
IUPAC Name 2-methylsulfanylacetonitrile
SMILES CSCC#N

Specifications

Density 1.052
Boiling Point 166°C to 167°C
Flash Point 67°C (152°F)
Refractive Index 1.482
Linear Formula CH3SCH2CN
Quantity 100 g
Beilstein 1738015
Formula Weight 87.14
Percent Purity 99%
Chemical Name or Material (Methylthio)acetonitrile
Hazard Category H227-H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 252-383-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only